Order Entry
Puerto Rico
ContactUsLinkComponent
 
Anti-WDR35 Rabbit Recombinant Polyclonal Antibody
New Product
 
undefined
Anti-WDR35 Rabbit Recombinant Polyclonal Antibody
New Product

 

  • Catalog No:
  • 77840-128
  • 77840-127
  • Antigen Symbol:
  • WDR35
  • WDR35
  • Antigen Name:
  • WD repeat-containing protein 35 protein (954-1181AA)
  • WD repeat-containing protein 35 protein (954-1181AA)
  • Antigen Synonyms:
  • Intraflagellar transport protein 121 homolog|KIAA1336|MGC33196|Naofen|WD repeat domain 35|WD repeat-containing protein 35|WDR35|WDR35_HUMAN
  • Intraflagellar transport protein 121 homolog|KIAA1336|MGC33196|Naofen|WD repeat domain 35|WD repeat-containing protein 35|WDR35|WDR35_HUMAN
  • Clonality:
  • Recombinant Polyclonal
  • Recombinant Polyclonal
  • Conjugation:
  • Unconjugated
  • Unconjugated
  • Reactivity:
  • Human
  • Human
  • Host:
  • Rabbit
  • Rabbit
  • Gene ID:
  • Q9P2L0
  • Q9P2L0
  • Isotype:
  • IgG
  • IgG
  • ELISA:
  • Yes
  • Yes
  • ImmunoChemistry:
  • Yes
  • Yes
  • Form:
  • Liquid
  • Liquid
  • Purification:
  • >95%, Protein G purified
  • >95%, Protein G purified
  • Storage Buffer:
  • 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
  • 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
  • Storage Temperature:
  • Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
  • Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
  • Shipping Temperature:
  • Shipped on blue ice
  • Shipped on blue ice

 

Frequently Bought Together