Anti-NFAT5 Rabbit Polyclonal Antibody
77818-324
New Product
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- Antigen Name:NFAT5/TonEBP
- Antigen Symbol:NFAT5
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Mouse
- Western Blot:Yes
- Size:100 µl
- Environmentally Preferable:
- Form:Liquid
- Gene ID:O94916
- Antigen Synonyms:Glutamine rich protein H65| KIAA0827| NF AT5| NF-AT5| NFAT 5| NFAT L1| NFAT like protein 1| NFAT5| NFAT5_HUMAN| NFATL 1| NFATL1| NFATZ| Nuclear factor of activated T cells 5| Nuclear factor of activated T cells 5 tonicity responsive| Nuclear factor of activated T cells| Nuclear factor of activated T-cells 5| OREBP| Osmotic response element binding protein| T cell transcription factor NFAT 5| T cell transcription factor NFAT5| T-cell transcription factor NFAT5| TonE binding protein| TonE-binding protein| TonEBP| Tonicity responsive enhancer binding protein| Tonicity-responsive enhancer-binding protein
- Storage Buffer:PBS, 150 mM NaCl, 0.02% sodium azide, 50% glycerol, pH 7.4
- Storage Temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on Blue ice
- Purification:Antigen affinity purified
- Packaging:Vial
Recommended dilution: WB: 1:500 to 1:3000
Type:
Antigen: NFAT5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse