Anti-SH3BGRL3 Rabbit Recombinant Antibody (Biotinylated)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: SH3BGRL3
Clonality: Recombinant
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77840-575
- 77840-576
- Antigen Symbol:
- SH3BGRL3
- SH3BGRL3
- Antigen Name:
- SH3 domain-binding glutamic acid-rich-like protein 3 protein (2-93AA)
- SH3 domain-binding glutamic acid-rich-like protein 3 protein (2-93AA)
- Antigen Synonyms:
- epididymis secretory protein Li 297|HEL-S-297|SH3 domain binding glutamic acid rich protein like 3|SH3 domain-binding glutamic acid-rich-like protein 3|SH3 domain-binding protein 1|Sh3bgrl3|SH3BGRL3 like protein|SH3BP-1|SH3L3_HUMAN|TIP B1|TIP-B1|TNF inhibitory protein
- epididymis secretory protein Li 297|HEL-S-297|SH3 domain binding glutamic acid rich protein like 3|SH3 domain-binding glutamic acid-rich-like protein 3|SH3 domain-binding protein 1|Sh3bgrl3|SH3BGRL3 like protein|SH3BP-1|SH3L3_HUMAN|TIP B1|TIP-B1|TNF inhibitory protein
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9H299
- Q9H299
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice