Anti-ACTL8 Rabbit Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: ELISA: 1:2000 to 1:5000, WB: 1:500 to 1:2000, IHC: 1:15 to 1:50
Type:
Antigen: ACTL8
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77825-724
- 77825-725
- Antigen Symbol:
- ACTL8
- ACTL8
- Antigen Name:
- Fusion protein of ACTL8
- Fusion protein of ACTL8
- Antigen Synonyms:
- Actin like 8|Actin like protein 8|Actin like protein|Actin-like protein 8|ACTL8|ACTL8_HUMAN|Cancer/testis antigen 57|CT57
- Actin like 8|Actin like protein 8|Actin like protein|Actin-like protein 8|ACTL8|ACTL8_HUMAN|Cancer/testis antigen 57|CT57
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9H568
- Q9H568
- Isotype:
- IgG
- IgG
- Western Blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice