Anti-FERMT3 Rabbit Recombinant Polyclonal Antibody (HRP)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: FERMT3
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77832-206
- 77832-207
- Antigen Symbol:
- FERMT3
- FERMT3
- Antigen Name:
- Fermitin family homolog 3 protein (388-503AA)
- Fermitin family homolog 3 protein (388-503AA)
- Antigen Synonyms:
- Fermitin family homolog 3|Fermitin family member 3|FERMT3|Kind3|Kindlin 3|Kindlin-3|Kindlin3|MGC10966|MIG 2|MIG2 like protein|MIG2-like protein|MIG2B|Unc 112 related protein 2|Unc-112-related protein 2|UNC112C|URP2|URP2_HUMAN|URP2SF
- Fermitin family homolog 3|Fermitin family member 3|FERMT3|Kind3|Kindlin 3|Kindlin-3|Kindlin3|MGC10966|MIG 2|MIG2 like protein|MIG2-like protein|MIG2B|Unc 112 related protein 2|Unc-112-related protein 2|UNC112C|URP2|URP2_HUMAN|URP2SF
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- HRP (Horseradish Peroxidase)
- HRP (Horseradish Peroxidase)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q86UX7
- Q86UX7
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together