Anti-RAB11FIP5 Rabbit Recombinant Polyclonal Antibody (FITC)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: RAB11FIP5
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77835-917
- 77835-916
- Antigen Symbol:
- RAB11FIP5
- RAB11FIP5
- Antigen Name:
- Rab11 family-interacting protein 5 protein (374-545AA)
- Rab11 family-interacting protein 5 protein (374-545AA)
- Antigen Synonyms:
- GAF 1|Gaf-1|GAF1|gamma SNAP associated factor 1|Gamma-SNAP-associated factor 1|KIAA0857|Phosphoprotein pp75|pp75|RAB11 family interacting protein 5 (class I)|Rab11 family-interacting protein 5|Rab11 FIP5|Rab11 interacting protein|Rab11 interacting protein Rip11|Rab11-FIP5|Rab11-interacting protein Rip11|Rab11fip5|RFIP5_HUMAN|RIP11
- GAF 1|Gaf-1|GAF1|gamma SNAP associated factor 1|Gamma-SNAP-associated factor 1|KIAA0857|Phosphoprotein pp75|pp75|RAB11 family interacting protein 5 (class I)|Rab11 family-interacting protein 5|Rab11 FIP5|Rab11 interacting protein|Rab11 interacting protein Rip11|Rab11-FIP5|Rab11-interacting protein Rip11|Rab11fip5|RFIP5_HUMAN|RIP11
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9BXF6
- Q9BXF6
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, protein G purified
- >95%, protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice