Anti-MTAP Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: MTAP
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77824-723
- 77824-722
- Antigen Symbol:
- MTAP
- MTAP
- Antigen Name:
- S-methyl-5\'-thioadenosine phosphorylase protein (1-283AA)
- S-methyl-5\'-thioadenosine phosphorylase protein (1-283AA)
- Antigen Synonyms:
- 5' methylthioadenosine phosphorylase|5''-methylthioadenosine phosphorylase|BDMF|c86fus|DMSFH|DMSMFH|Epididymis luminal protein 249|HEL 249|LGMBF|MeSAdo phosphorylase|Methylthioadenosine phosphorylase|MSAP|MTA phosphorylase|MTAP|MTAP_HUMAN|MTAPase|S methyl 5 thioadenosine phosphorylase|S methyl 5' thioadenosine phosphorylase|S-methyl-5''-thioadenosine phosphorylase
- 5' methylthioadenosine phosphorylase|5''-methylthioadenosine phosphorylase|BDMF|c86fus|DMSFH|DMSMFH|Epididymis luminal protein 249|HEL 249|LGMBF|MeSAdo phosphorylase|Methylthioadenosine phosphorylase|MSAP|MTA phosphorylase|MTAP|MTAP_HUMAN|MTAPase|S methyl 5 thioadenosine phosphorylase|S methyl 5' thioadenosine phosphorylase|S-methyl-5''-thioadenosine phosphorylase
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q13126
- Q13126
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice