Anti-TRDN Rabbit Recombinant Polyclonal Antibody (Biotin)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: TRDN
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77811-495
- 77803-495
- Antigen Symbol:
- TRDN
- TRDN
- Antigen Name:
- Triadin protein (75-161AA)
- Triadin protein (75-161AA)
- Antigen Synonyms:
- dJ166D18.1|DKFZp779I2253|MGC88285|OTTHUMP00000040354|TDN|TRDN|TRDN_HUMAN|Triadin|TRISK 51|TRISK|TRISK51
- dJ166D18.1|DKFZp779I2253|MGC88285|OTTHUMP00000040354|TDN|TRDN|TRDN_HUMAN|Triadin|TRISK 51|TRISK|TRISK51
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q13061
- Q13061
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice