Anti-SCN3A Rabbit Recombinant Polyclonal Antibody
New Product
Recommended dilution: IHC: 1:20 to 1:200
Type:
Antigen: SCN3A
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77842-507
- 77842-506
- Antigen Symbol:
- SCN3A
- SCN3A
- Antigen Name:
- Sodium channel protein type 3 subunit alpha protein (274-400AA)
- Sodium channel protein type 3 subunit alpha protein (274-400AA)
- Antigen Synonyms:
- brain III voltage-gated sodium channel|NAC3|Nav1.3|SCN3A_HUMAN|Sodium channel protein brain III subunit alpha|Sodium channel protein type 3 subunit alpha|Sodium channel protein type III subunit alpha|Sodium channel protein, brain III subunit alpha|sodium channel, brain type III, alpha subunit|sodium channel, neuronal type III, alpha subunit|sodium channel, voltage-gated, type III, alpha polypeptide|sodium channel, voltage-gated, type III, alpha subunit|Voltage gated sodium channel subtype III|Voltage gated sodium channel subunit alpha Nav1.3|Voltage-gated sodium channel subtype III|Voltage-gated sodium channel subunit alpha Nav1.3
- brain III voltage-gated sodium channel|NAC3|Nav1.3|SCN3A_HUMAN|Sodium channel protein brain III subunit alpha|Sodium channel protein type 3 subunit alpha|Sodium channel protein type III subunit alpha|Sodium channel protein, brain III subunit alpha|sodium channel, brain type III, alpha subunit|sodium channel, neuronal type III, alpha subunit|sodium channel, voltage-gated, type III, alpha polypeptide|sodium channel, voltage-gated, type III, alpha subunit|Voltage gated sodium channel subtype III|Voltage gated sodium channel subunit alpha Nav1.3|Voltage-gated sodium channel subtype III|Voltage-gated sodium channel subunit alpha Nav1.3
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9NY46
- Q9NY46
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice