Anti-DYNLRB1 Rabbit Recombinant Polyclonal Antibody (FITC)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: DYNLRB1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77836-382
- 77836-381
- Antigen Symbol:
- DYNLRB1
- DYNLRB1
- Antigen Name:
- Dynein light chain roadblock-type 1 protein (3-96AA)
- Dynein light chain roadblock-type 1 protein (3-96AA)
- Antigen Synonyms:
- DYNLRB1|BITH|DNCL2A|DNLC2A|ROBLD1|HSPC162Dynein light chain roadblock-type 1|Bithoraxoid-like protein|BLP|Dynein light chain 2A|cytoplasmic|Dynein-associated protein Km23|Roadblock domain-containing protein 1
- DYNLRB1|BITH|DNCL2A|DNLC2A|ROBLD1|HSPC162Dynein light chain roadblock-type 1|Bithoraxoid-like protein|BLP|Dynein light chain 2A|cytoplasmic|Dynein-associated protein Km23|Roadblock domain-containing protein 1
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9NP97
- Q9NP97
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, protein G purified
- >95%, protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together