Anti-ZNF423 Rabbit Polyclonal Antibody (FITC)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: ZNF423
Clonality: Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77805-441
- 77805-440
- Antigen Symbol:
- ZNF423
- ZNF423
- Antigen Name:
- Zinc finger protein 423 protein (39-57AA)
- Zinc finger protein 423 protein (39-57AA)
- Antigen Synonyms:
- Early B cell factor associated zinc finger protein|Ebfaz|hOAZ|JBTS19|KIAA0760|NPHP14|Nur12|OAZ|OLF 1/EBF associated zinc finger|OLF1/EBF associated zinc finger protein|Olf1/EBF-associated zinc finger protein|Roaz|Smad and Olf interacting zinc finger protein|Smad- and Olf-interacting zinc finger protein|Zfp104|ZFP423|zinc finger protein 423, mouse, homolog of|Zinc finger protein 423|ZN423_HUMAN|ZNF423|ZNF423 zinc finger protein 423
- Early B cell factor associated zinc finger protein|Ebfaz|hOAZ|JBTS19|KIAA0760|NPHP14|Nur12|OAZ|OLF 1/EBF associated zinc finger|OLF1/EBF associated zinc finger protein|Olf1/EBF-associated zinc finger protein|Roaz|Smad and Olf interacting zinc finger protein|Smad- and Olf-interacting zinc finger protein|Zfp104|ZFP423|zinc finger protein 423, mouse, homolog of|Zinc finger protein 423|ZN423_HUMAN|ZNF423|ZNF423 zinc finger protein 423
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q2M1K9
- Q2M1K9
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice