Anti-MTMR3 Rabbit Recombinant Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: IHC: 1:200 to 1:500, IF: 1:50 to 1:200
Type:
Antigen: MTMR3
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77824-781
- 77824-780
- Antigen Symbol:
- MTMR3
- MTMR3
- Antigen Name:
- Myotubularin-related protein 3 protein (652-899AA)
- Myotubularin-related protein 3 protein (652-899AA)
- Antigen Synonyms:
- FYVE domain-containing dual specificity protein phosphatase 1|FYVE-DSP1|MTMR3|MTMR3_HUMAN|Myotubularin-related protein 3|Zinc finger FYVE domain-containing protein 10
- FYVE domain-containing dual specificity protein phosphatase 1|FYVE-DSP1|MTMR3|MTMR3_HUMAN|Myotubularin-related protein 3|Zinc finger FYVE domain-containing protein 10
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q13615
- Q13615
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice