Anti-USP31 Rabbit Recombinant Polyclonal Antibody (FITC)
New Product
Type:
Antigen: USP31
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77830-185
- 77830-186
- Antigen Symbol:
- USP31
- USP31
- Antigen Name:
- Ubiquitin carboxyl-terminal hydrolase 31 protein (1069-1196AA)
- Ubiquitin carboxyl-terminal hydrolase 31 protein (1069-1196AA)
- Antigen Synonyms:
- Deubiquitinating enzyme 31| KIAA1203| Ubiquitin carboxyl terminal hydrolase 31| Ubiquitin carboxyl-terminal hydrolase 31| Ubiquitin specific peptidase 31| Ubiquitin specific processing protease 31| Ubiquitin specific protease 31| Ubiquitin specific proteinase 31| Ubiquitin thioesterase 31| Ubiquitin thiolesterase 31| Ubiquitin-specific-processing protease 31| UBP31_HUMAN| USP 31| USP31
- Deubiquitinating enzyme 31| KIAA1203| Ubiquitin carboxyl terminal hydrolase 31| Ubiquitin carboxyl-terminal hydrolase 31| Ubiquitin specific peptidase 31| Ubiquitin specific processing protease 31| Ubiquitin specific protease 31| Ubiquitin specific proteinase 31| Ubiquitin thioesterase 31| Ubiquitin thiolesterase 31| Ubiquitin-specific-processing protease 31| UBP31_HUMAN| USP 31| USP31
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q70CQ4
- Q70CQ4
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on Blue ice
- Shipped on Blue ice