Anti-MAP2K5 Rabbit Recombinant Antibody (HRP (Horseradish Peroxidase))
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: MAP2K5
Clonality: Recombinant
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77822-847
- 77822-848
- Antigen Symbol:
- MAP2K5
- MAP2K5
- Antigen Name:
- Dual specificity mitogen-activated protein kinase kinase 5 protein (1-190AA)
- Dual specificity mitogen-activated protein kinase kinase 5 protein (1-190AA)
- Antigen Synonyms:
- Dual specificity mitogen activated protein kinase kinase 5|Dual specificity mitogen-activated protein kinase kinase 5|EC 2.7.12.2|HsT17454|MAP kinase kinase 5|MAP kinase kinase MEK5b|MAP2K5|MAPK/ERK kinase 5|MAPKK 5|MAPKK5|MEK 5|mitogen-activated protein kinase kinase 5|MKK5|MP2K5_HUMAN|PRKMK5|Protein kinase, mitogen-activated, kinase 5|SAPKK5|SKK5
- Dual specificity mitogen activated protein kinase kinase 5|Dual specificity mitogen-activated protein kinase kinase 5|EC 2.7.12.2|HsT17454|MAP kinase kinase 5|MAP kinase kinase MEK5b|MAP2K5|MAPK/ERK kinase 5|MAPKK 5|MAPKK5|MEK 5|mitogen-activated protein kinase kinase 5|MKK5|MP2K5_HUMAN|PRKMK5|Protein kinase, mitogen-activated, kinase 5|SAPKK5|SKK5
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- HRP (Horseradish Peroxidase)
- HRP (Horseradish Peroxidase)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q13163
- Q13163
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice