Anti-SCP Rabbit Recombinant Polyclonal Antibody (FITC)
New Product
Type:
Antigen: SCP
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Epstein-Barr virus
- Catalog No:
- 77815-283
- 77815-284
- Antigen Symbol:
- SCP
- SCP
- Antigen Name:
- Recombinant Epstein-Barr virus Small capsomere-interacting protein (31-176AA)
- Recombinant Epstein-Barr virus Small capsomere-interacting protein (31-176AA)
- Antigen Synonyms:
- SCP|BFRF3|Small capsomere-interacting protein
- SCP|BFRF3|Small capsomere-interacting protein
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- EBV (Epstein-Barr virus)
- EBV (Epstein-Barr virus)
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P14348
- P14348
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, protein G purified
- >95%, protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice