Anti-CYB5R1 Rabbit Polyclonal Antibody
77818-585
New Product
- Antigen Name:Synthesized peptide derived from internal of CYB5R1
- Antigen Symbol:CYB5R1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Mouse,Rat
- Western Blot:Yes
- Size:100 µl
- Form:Liquid
- Gene ID:Q9UHQ9
- Antigen Synonyms:b5R.1|B5R1|B5R2|CYB5R1|Cytochrome b5 reductase 1|EC 1.6.2.2|Humb5R2|NAD|NAD(P)H:quinone oxidoreductase type 3 polypeptide A2|NAD(P)H:quinone oxidoreductase type 3, polypeptide A2|NADH cytochrome b5 reductase 1|NADH-cytochrome b5 reductase 1|NB5R1_HUMAN|NQO3A2|UNQ3049/PRO9865
- Storage Buffer:PBS, 150 mM NaCl, 0.02% sodium azide, 50% glycerol, pH 7.4
- Storage Temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Purification:Antigen affinity purified
- Packaging:Vial
Recommended dilution: WB: 1:500 to 1:3000
Type:
Antigen: CYB5R1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat