Anti-HSPA14 Rabbit Polyclonal Antibody
Catalog # 77797-994
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Name:Synthesized peptide derived from the C-terminal region of HSP60.
- Antigen Symbol:HSPA14
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Mouse,Rat
- Western Blot:Yes
- Size:50 µg
- Form:Liquid
- Gene ID:Q0VDF9
- Antigen Synonyms:Heat shock 70 kDa protein 14| Heat shock 70kDa protein 14| Heat shock protein HSP60| Heat shock protein hsp70 related protein| HSP60| HSP70 4| HSP70 like protein 1| HSP70-like protein 1| HSP70L1| HSP7E_HUMAN| HSPA 14| HSPA14| MGC131990
- Storage Buffer:PBS, 50% glycerol, 0.5% BSA, 0.02% sodium azide, pH 7.4
- Storage Temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Purification:Antigen affinity purified
- Packaging:Vial
Specifications
About this item
Recommended dilution: WB: 1:500 to 1:2000, IHC: 1:100 to 1:300, ELISA: 1:40000
Type:
Antigen: HSPA14
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat