Anti-YME1L1 Rabbit Recombinant Polyclonal Antibody
77833-270
New Product
- Antigen Name:ATP-dependent zinc metalloprotease YME1L1 protein (1-240AA)
- Antigen Symbol:YME1L1
- Clonality:Recombinant Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:50 µl
- Form:Liquid
- Gene ID:Q96TA2
- Antigen Synonyms:ATP dependent metalloprotease FtsH1 homolog| ATP-dependent metalloprotease FtsH1| ATP-dependent metalloprotease YME1L1| ATP-dependent zinc metalloprotease YME1L1| FTSH1| Meg-4| MEG4| PAMP| Presenilin-associated metalloprotease| UNQ1868/PRO4304| YME1 like 1| YME1-like protein 1| YME1L| Yme1l1| YMEL1_HUMAN
- Storage Buffer:PBS, 0.02% sodium azide, 50% glycerol, pH 7.3
- Storage Temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Purification:Antigen affinity purified
- Packaging:Vial
Recommended dilution: WB: 1:1000 to 1:5000, IHC: 1:20 to 1:200
Type:
Antigen: YME1L1
Clonality: Recombinant Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human