Anti-ACOX1 Rabbit Recombinant Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: IHC: 1:20 to 1:200, IF: 1:50 to 1:200
Type:
Antigen: ACOX1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77823-158
- 77823-159
- Antigen Symbol:
- ACOX1
- ACOX1
- Antigen Name:
- Peroxisomal acyl-coenzyme A oxidase 1 protein (113-273AA)
- Peroxisomal acyl-coenzyme A oxidase 1 protein (113-273AA)
- Antigen Synonyms:
- ACOX|ACOX1|ACOX1_HUMAN|Acyl CoA oxidase 1 palmitoyl|Acyl CoA oxidase straight chain|AOX|EC 1.3.3.6|PALMCOX|Palmitoyl CoA oxidase|Palmitoyl-CoA oxidase|Peroxisomal acyl coenzyme A oxidase 1|Peroxisomal acyl-coenzyme A oxidase 1|Peroxisomal fatty acyl CoA oxidase|SCOX|Straight chain acyl CoA oxidase|Straight-chain acyl-CoA oxidase
- ACOX|ACOX1|ACOX1_HUMAN|Acyl CoA oxidase 1 palmitoyl|Acyl CoA oxidase straight chain|AOX|EC 1.3.3.6|PALMCOX|Palmitoyl CoA oxidase|Palmitoyl-CoA oxidase|Peroxisomal acyl coenzyme A oxidase 1|Peroxisomal acyl-coenzyme A oxidase 1|Peroxisomal fatty acyl CoA oxidase|SCOX|Straight chain acyl CoA oxidase|Straight-chain acyl-CoA oxidase
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q15067
- Q15067
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice