Anti-AKAP13 Rabbit Recombinant Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: IHC: 1:20 to 1:200
Type:
Antigen: AKAP13
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77821-778
- 77821-777
- Antigen Symbol:
- AKAP13
- AKAP13
- Antigen Name:
- A-kinase anchor protein 13 protein (1-150AA)
- A-kinase anchor protein 13 protein (1-150AA)
- Antigen Synonyms:
- A kinase (PRKA) anchor protein 13|A kinase anchor protein 13|A kinase anchoring protein|A-kinase anchor protein 13|AKAP 13|AKAP Lbc|AKAP-13|AKAP-Lbc|AKP13_HUMAN|ARHGEF13|Breast cancer nuclear receptor binding auxiliary protein|Breast cancer nuclear receptor-binding auxiliary protein|BRX|c lbc|FLJ11952|FLJ43341|Guanine nucleotide exchange factor Lbc|HA 3|HA3|Ht 31|Ht31|Human thyroid anchoring protein 31|Human thyroid-anchoring protein 31|LBC|LBC oncogene|Lymphoid blast crisis oncogene|Non oncogenic Rho GTPase specific GTP exchange factor|Non-oncogenic Rho GTPase-specific GTP exchange factor|p47|Protein kinase A anchoring protein 13|Protein kinase A-anchoring protein 13|PROTO LB|PROTO LBC
- A kinase (PRKA) anchor protein 13|A kinase anchor protein 13|A kinase anchoring protein|A-kinase anchor protein 13|AKAP 13|AKAP Lbc|AKAP-13|AKAP-Lbc|AKP13_HUMAN|ARHGEF13|Breast cancer nuclear receptor binding auxiliary protein|Breast cancer nuclear receptor-binding auxiliary protein|BRX|c lbc|FLJ11952|FLJ43341|Guanine nucleotide exchange factor Lbc|HA 3|HA3|Ht 31|Ht31|Human thyroid anchoring protein 31|Human thyroid-anchoring protein 31|LBC|LBC oncogene|Lymphoid blast crisis oncogene|Non oncogenic Rho GTPase specific GTP exchange factor|Non-oncogenic Rho GTPase-specific GTP exchange factor|p47|Protein kinase A anchoring protein 13|Protein kinase A-anchoring protein 13|PROTO LB|PROTO LBC
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q12802
- Q12802
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice