Anti-ZNF2 Rabbit Recombinant Polyclonal Antibody (FITC)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: ZNF2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77805-112
- 77805-113
- Antigen Symbol:
- ZNF2
- ZNF2
- Antigen Name:
- Zinc finger protein 2 protein (134-289AA)
- Zinc finger protein 2 protein (134-289AA)
- Antigen Synonyms:
- A1 5|A1-5|Zfp661|Zinc finger protein 2|Zinc finger protein 2.2|Zinc finger protein 661|ZNF 2|ZNF2|ZNF2_HUMAN|ZNF661
- A1 5|A1-5|Zfp661|Zinc finger protein 2|Zinc finger protein 2.2|Zinc finger protein 661|ZNF 2|ZNF2|ZNF2_HUMAN|ZNF661
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9BSG1
- Q9BSG1
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:
- Upon receipt, store at −20...−80 °C. Avoid repeated freeze.
- Upon receipt, store at −20...−80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice