Anti-PYCR1 Rabbit Recombinant Polyclonal Antibody (HRP)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: PYCR1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77796-252
- 77796-251
- Antigen Symbol:
- PYCR1
- PYCR1
- Antigen Name:
- Pyrroline-5-carboxylate reductase 1, mitochondrial protein (2-319AA)
- Pyrroline-5-carboxylate reductase 1, mitochondrial protein (2-319AA)
- Antigen Synonyms:
- PYCR1|Pyrroline-5-carboxylate reductase 1|mitochondrial|P5C reductase 1|P5CR 1|EC 1.5.1.2
- PYCR1|Pyrroline-5-carboxylate reductase 1|mitochondrial|P5C reductase 1|P5CR 1|EC 1.5.1.2
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- HRP (Horseradish Peroxidase)
- HRP (Horseradish Peroxidase)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P32322
- P32322
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice