Anti-CAPNS1 Rabbit Recombinant Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: IF:1:500 to 1:1000
Type:
Antigen: CAPNS1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77806-549
- 77806-548
- Antigen Symbol:
- CAPNS1
- CAPNS1
- Antigen Name:
- Calpain small subunit 1 protein (1-268AA)
- Calpain small subunit 1 protein (1-268AA)
- Antigen Synonyms:
- 30K|Calcium activated neutral proteinase|Calcium activated neutral proteinase small subunit|Calcium dependent protease small subunit 1|Calcium dependent protease small subunit|Calcium-activated neutral proteinase small subunit|Calcium-dependent protease small subunit 1|Calcium-dependent protease small subunit|Calpain 4|Calpain 4 small subunit (30K)|Calpain 4 small subunit|Calpain regulatory subunit|Calpain S1|Calpain small polypeptide|Calpain small subunit 1|Calpain4|CalpainS1|CANP|CANP small subunit|CANPS|CAPN 4|CAPN S1|CAPN4|CAPNS 1|CAPNS|Capns1|CDPS|CPNS1_HUMAN|CSS 1|CSS1
- 30K|Calcium activated neutral proteinase|Calcium activated neutral proteinase small subunit|Calcium dependent protease small subunit 1|Calcium dependent protease small subunit|Calcium-activated neutral proteinase small subunit|Calcium-dependent protease small subunit 1|Calcium-dependent protease small subunit|Calpain 4|Calpain 4 small subunit (30K)|Calpain 4 small subunit|Calpain regulatory subunit|Calpain S1|Calpain small polypeptide|Calpain small subunit 1|Calpain4|CalpainS1|CANP|CANP small subunit|CANPS|CAPN 4|CAPN S1|CAPN4|CAPNS 1|CAPNS|Capns1|CDPS|CPNS1_HUMAN|CSS 1|CSS1
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P04632
- P04632
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice