Anti-PRKAB2 Rabbit Recombinant Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: IHC: 1:20 to 1:200
Type:
Antigen: PRKAB2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77819-440
- 77819-441
- Antigen Symbol:
- PRKAB2
- PRKAB2
- Antigen Name:
- 5\\\'-AMP-activated protein kinase subunit beta-2 protein (1-272AA)
- 5\\\'-AMP-activated protein kinase subunit beta-2 protein (1-272AA)
- Antigen Synonyms:
- 5' AMP activated protein kinase beta 2 subunit|5' AMP activated protein kinase subunit beta 2|5''-AMP-activated protein kinase subunit beta-2|AAKB2_HUMAN|AMP activated protein kinase beta 2 non catalytic subunit|AMPK beta 2|AMPK beta 2 chain|AMPK subunit beta 2|AMPK subunit beta-2|MGC61468|PRKAB 2|Protein kinase AMP activated beta 2 non catalytic subunit
- 5' AMP activated protein kinase beta 2 subunit|5' AMP activated protein kinase subunit beta 2|5''-AMP-activated protein kinase subunit beta-2|AAKB2_HUMAN|AMP activated protein kinase beta 2 non catalytic subunit|AMPK beta 2|AMPK beta 2 chain|AMPK subunit beta 2|AMPK subunit beta-2|MGC61468|PRKAB 2|Protein kinase AMP activated beta 2 non catalytic subunit
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- O43741
- O43741
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3.
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3.
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on Blue ice
- Shipped on Blue ice