Anti-SMC2 Rabbit Recombinant Polyclonal Antibody
New Product
Recommended dilution: IF: 1:50 to 1:200
Type:
Antigen: SMC2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77800-314
- 77800-315
- Antigen Symbol:
- SMC2
- SMC2
- Antigen Name:
- Structural maintenance of chromosomes protein 2 protein (861-968AA)
- Structural maintenance of chromosomes protein 2 protein (861-968AA)
- Antigen Synonyms:
- CAP E|CAPE|Chromosome associated protein E|Chromosome-associated protein E|FLJ10093|hCAP E|hCAP-E|hCAPE|PRO0324|SMC 2|SMC protein 2|SMC-2|SMC2|SMC2 L1|SMC2_HUMAN|SMC2L1|Structural maintenance of chromosomes (SMC) family member chromosome associated protein E|Structural maintenance of chromosomes 2|Structural maintenance of chromosomes 2 like 1|Structural maintenance of chromosomes 2 yeast like 1|Structural maintenance of chromosomes protein 2|XCAP E homolog|XCAP-E homolog|XCAPE homolog
- CAP E|CAPE|Chromosome associated protein E|Chromosome-associated protein E|FLJ10093|hCAP E|hCAP-E|hCAPE|PRO0324|SMC 2|SMC protein 2|SMC-2|SMC2|SMC2 L1|SMC2_HUMAN|SMC2L1|Structural maintenance of chromosomes (SMC) family member chromosome associated protein E|Structural maintenance of chromosomes 2|Structural maintenance of chromosomes 2 like 1|Structural maintenance of chromosomes 2 yeast like 1|Structural maintenance of chromosomes protein 2|XCAP E homolog|XCAP-E homolog|XCAPE homolog
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- O95347
- O95347
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice