Anti-VARS2 Rabbit Recombinant Polyclonal Antibody (Biotinylated)
New Product
Type:
Antigen: VARS2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77829-132
- 77829-131
- Antigen Symbol:
- VARS2
- VARS2
- Antigen Name:
- Valine--tRNA ligase, mitochondrial protein (830-981AA)
- Valine--tRNA ligase, mitochondrial protein (830-981AA)
- Antigen Synonyms:
- COXPD20|mitochondrial|SYVM_HUMAN|Valine--tRNA ligase|ValRS|Valyl tRNA synthetase 2 mitochondrial|Valyl-tRNA synthetase|Valyl-tRNA synthetase-like|VARS2L|VARSL
- COXPD20|mitochondrial|SYVM_HUMAN|Valine--tRNA ligase|ValRS|Valyl tRNA synthetase 2 mitochondrial|Valyl-tRNA synthetase|Valyl-tRNA synthetase-like|VARS2L|VARSL
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q5ST30
- Q5ST30
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice