Anti-FHL5 Rabbit Recombinant Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: IF: 1:50 to 1:200
Type:
Antigen: FHL5
Clonality: Recombinant
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77828-597
- 77828-598
- Antigen Symbol:
- FHL5
- FHL5
- Antigen Name:
- Four and a half LIM domains protein 5 protein (16-284AA)
- Four and a half LIM domains protein 5 protein (16-284AA)
- Antigen Synonyms:
- Activator of cAMP responsive element modulator (CREM) in testis|Activator of cAMP-responsive element modulator in testis|Activator of CREM in testis|FHL-5|Fhl5|FHL5 protein|FHL5_HUMAN|Four and a half LIM domains protein 5
- Activator of cAMP responsive element modulator (CREM) in testis|Activator of cAMP-responsive element modulator in testis|Activator of CREM in testis|FHL-5|Fhl5|FHL5 protein|FHL5_HUMAN|Four and a half LIM domains protein 5
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q5TD97
- Q5TD97
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice