Anti-DYNLRB1 Rabbit Recombinant Polyclonal Antibody (Biotin)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: DYNLRB1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77836-384
- 77836-383
- Antigen Symbol:
- DYNLRB1;BITH;DNCL2A;DNLC2A;ROBLD1;HSPC162Dynein light chain roadblock-type 1;Bithoraxoid-like protein;BLP;Dynein light chain 2A;cytoplasmic;Dynein-associated protein Km23;Roadblock domain-containing protein 1
- DYNLRB1;BITH;DNCL2A;DNLC2A;ROBLD1;HSPC162Dynein light chain roadblock-type 1;Bithoraxoid-like protein;BLP;Dynein light chain 2A;cytoplasmic;Dynein-associated protein Km23;Roadblock domain-containing protein 1
- Antigen Name:
- Dynein light chain roadblock-type 1 protein (3-96AA)
- Dynein light chain roadblock-type 1 protein (3-96AA)
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9NP97
- Q9NP97
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice