Anti-PPP6C Rabbit Recombinant Polyclonal Antibody (Biotinylated)
New Product
Type:
Antigen: PPP6C
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77795-155
- 77795-156
- Antigen Symbol:
- PPP6C
- PPP6C
- Antigen Name:
- Serine/threonine-protein phosphatase 6 catalytic subunit protein (40-211AA)
- Serine/threonine-protein phosphatase 6 catalytic subunit protein (40-211AA)
- Antigen Synonyms:
- FLJ92648|MGC12249|PP 6|PP6|PP6C|PPP 6|PPP 6C|PPP6|PPP6_HUMAN|Ppp6c|Protein phosphatase 6 catalytic subunit|Serine/threonine protein phosphatase 6|Serine/threonine protein phosphatase catalytic subunit|Serine/threonine-protein phosphatase 6 catalytic subunit
- FLJ92648|MGC12249|PP 6|PP6|PP6C|PPP 6|PPP 6C|PPP6|PPP6_HUMAN|Ppp6c|Protein phosphatase 6 catalytic subunit|Serine/threonine protein phosphatase 6|Serine/threonine protein phosphatase catalytic subunit|Serine/threonine-protein phosphatase 6 catalytic subunit
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- O00743
- O00743
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice