Anti-UGT2A3 Rabbit Recombinant Polyclonal Antibody (HRP)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: UGT2A3
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77831-184
- 77831-185
- Antigen Symbol:
- UGT2A3
- UGT2A3
- Antigen Name:
- UDP-glucuronosyltransferase 2A3 protein (201-491AA)
- UDP-glucuronosyltransferase 2A3 protein (201-491AA)
- Antigen Synonyms:
- UGT2A3|UNQ2559/PRO6239|UDP-glucuronosyltransferase 2A3|UDPGT 2A3|EC 2.4.1.17
- UGT2A3|UNQ2559/PRO6239|UDP-glucuronosyltransferase 2A3|UDPGT 2A3|EC 2.4.1.17
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- HRP (Horseradish Peroxidase)
- HRP (Horseradish Peroxidase)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q6UWM9
- Q6UWM9
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice