Anti-CBX6 Rabbit Polyclonal Antibody
Catalog # 77813-618
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Name:Synthesized peptide derived from internal of CBX6
- Antigen Symbol:CBX6
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Mouse,Rat
- Western Blot:Yes
- Size:100 µl
- Form:Liquid
- Gene ID:O95503
- Antigen Synonyms:Cbx 6|Cbx6|CBX6_HUMAN|Chromobox homolog 6|Chromobox protein homolog 6
- Storage Buffer:PBS, 150 mM NaCl, 0.02% Sodium azide, 50% glycerol, pH 7.4
- Storage Temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Purification:Antigen affinity purified
- Packaging:Vial
Specifications
About this item
Recommended dilution: WB: 1:500 to 1:3000
Type:
Antigen: CBX6
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat