Anti-PDE11A Rabbit Recombinant Polyclonal Antibody (HRP)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: PDE11A
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77839-582
- 77839-581
- Antigen Symbol:
- PDE11A
- PDE11A
- Antigen Name:
- Dual 3\',5\'-cyclic-AMP and -GMP phosphodiesterase 11A protein (42-190AA)
- Dual 3\',5\'-cyclic-AMP and -GMP phosphodiesterase 11A protein (42-190AA)
- Antigen Synonyms:
- 5''-cyclic-AMP and -GMP phosphodiesterase 11A|cAMP and cGMP phosphodiesterase 11A|cGMP phosphodiesterase 11A|Cyclic nucleotide phosphodiesterase 11A|Dual 3''|Dual 3',5'-cyclic-AMP and -GMP phosphodiesterase 11A|pde11|PDE11_HUMAN|pde11a|PDE11A1|pde11a2|Phosphodiesterase 11A|PPNAD2
- 5''-cyclic-AMP and -GMP phosphodiesterase 11A|cAMP and cGMP phosphodiesterase 11A|cGMP phosphodiesterase 11A|Cyclic nucleotide phosphodiesterase 11A|Dual 3''|Dual 3',5'-cyclic-AMP and -GMP phosphodiesterase 11A|pde11|PDE11_HUMAN|pde11a|PDE11A1|pde11a2|Phosphodiesterase 11A|PPNAD2
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- HRP (Horseradish Peroxidase)
- HRP (Horseradish Peroxidase)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9HCR9
- Q9HCR9
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:
- Upon receipt, store at −20...−80 °C. Avoid repeated freeze.
- Upon receipt, store at −20...−80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice