Anti-ACOT6 Rabbit Recombinant Polyclonal Antibody
New Product
Recommended dilution: IHC: 1:20 to 1:200, IF: 1:50 to 1:200
Type:
Antigen: ACOT6
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77826-320
- 77826-321
- Antigen Symbol:
- ACOT6
- ACOT6
- Antigen Name:
- Putative acyl-coenzyme A thioesterase 6 protein (1-207AA)
- Putative acyl-coenzyme A thioesterase 6 protein (1-207AA)
- Antigen Synonyms:
- ACOT6|C14orf42Putative acyl-coenzyme A thioesterase 6|Putative acyl-CoA thioesterase 6|EC 3.1.2.-
- ACOT6|C14orf42Putative acyl-coenzyme A thioesterase 6|Putative acyl-CoA thioesterase 6|EC 3.1.2.-
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q3I5F7
- Q3I5F7
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice