Order Entry
Puerto Rico
ContactUsLinkComponent
Anti-MAT2B Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Anti-MAT2B Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Supplier:  AFG Bioscience
New Product
Certificates
undefined
Anti-MAT2B Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Supplier:  AFG Bioscience
New Product

Specifications

  • Catalog No:
  • 77841-174
  • 77841-175
  • Antigen Symbol:
  • MAT2B
  • MAT2B
  • Antigen Name:
  • Methionine adenosyltransferase 2 subunit beta protein (85-143AA)
  • Methionine adenosyltransferase 2 subunit beta protein (85-143AA)
  • Antigen Synonyms:
  • 1110064C04Rik|2410018D16Rik|AI182287|AU022853|Beta regulatory subunit of methionine adenosyltransferase|dTDP 4 keto 6 deoxy D glucose 4 reductase|DTDP-4-keto-6-deoxy-D-glucose 4-reductase|MAT II|MAT II beta|Mat2b|MAT2B_HUMAN|MATIIbeta|Methionine adenosyltransferase 2 beta subunit|Methionine adenosyltransferase 2 subunit beta|Methionine adenosyltransferase II beta|MGC12237|MSTP045|Nbla02999|OTTMUSP00000005600|Putative dTDP-4-keto-6-deoxy-D-glucose 4-reductase|Putative protein product of Nbla02999|RP23-382C18.2|SDR23E1|Short chain dehydrogenase/reductase family 23E member 1|TGR
  • 1110064C04Rik|2410018D16Rik|AI182287|AU022853|Beta regulatory subunit of methionine adenosyltransferase|dTDP 4 keto 6 deoxy D glucose 4 reductase|DTDP-4-keto-6-deoxy-D-glucose 4-reductase|MAT II|MAT II beta|Mat2b|MAT2B_HUMAN|MATIIbeta|Methionine adenosyltransferase 2 beta subunit|Methionine adenosyltransferase 2 subunit beta|Methionine adenosyltransferase II beta|MGC12237|MSTP045|Nbla02999|OTTMUSP00000005600|Putative dTDP-4-keto-6-deoxy-D-glucose 4-reductase|Putative protein product of Nbla02999|RP23-382C18.2|SDR23E1|Short chain dehydrogenase/reductase family 23E member 1|TGR
  • Clonality:
  • Recombinant Polyclonal
  • Recombinant Polyclonal
  • Conjugation:
  • Biotin
  • Biotin
  • Reactivity:
  • Human
  • Human
  • Host:
  • Rabbit
  • Rabbit
  • Gene ID:
  • Q9NZL9
  • Q9NZL9
  • Isotype:
  • IgG
  • IgG
  • ELISA:
  • Yes
  • Yes
  • Form:
  • Liquid
  • Liquid
  • Purification:
  • >95%, Protein G purified
  • >95%, Protein G purified
  • Storage Buffer:
  • 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
  • 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
  • Storage Temperature:
  • Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
  • Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
  • Shipping Temperature:
  • Shipped on blue ice
  • Shipped on blue ice

Specifications