Anti-PF4 Rabbit Recombinant Polyclonal Antibody (HRP)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: PF4
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77793-821
- 77793-820
- Antigen Symbol:
- PF4
- PF4
- Antigen Name:
- Platelet factor 4 protein (32-101AA)
- Platelet factor 4 protein (32-101AA)
- Antigen Synonyms:
- C-X-C motif chemokine 4|Chemokine (C X C motif) ligand 4|Chemokine (CXC motif) ligand 4|chemokine ligand 4|CXCL 4|CXCL4|Iroplact|MGC138298|Oncostatin A|Oncostatin-A|OncostatinA|PF 4|PF-4|Pf4a|Platelet factor 4|PLF4_HUMAN|SCYB 4|SCYB4|short form|Small inducible cytokine subfamily B|Small inducible cytokine subfamily member 4
- C-X-C motif chemokine 4|Chemokine (C X C motif) ligand 4|Chemokine (CXC motif) ligand 4|chemokine ligand 4|CXCL 4|CXCL4|Iroplact|MGC138298|Oncostatin A|Oncostatin-A|OncostatinA|PF 4|PF-4|Pf4a|Platelet factor 4|PLF4_HUMAN|SCYB 4|SCYB4|short form|Small inducible cytokine subfamily B|Small inducible cytokine subfamily member 4
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- HRP (Horseradish Peroxidase)
- HRP (Horseradish Peroxidase)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P02776
- P02776
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on Blue ice
- Shipped on Blue ice