Anti-RABGEF1 Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: RABGEF1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77841-619
- 77841-620
- Antigen Symbol:
- RABGEF1
- RABGEF1
- Antigen Name:
- Rab5 GDP/GTP exchange factor protein (1-491AA)
- Rab5 GDP/GTP exchange factor protein (1-491AA)
- Antigen Synonyms:
- RAB guanine nucleotide exchange factor (GEF) 1|Rab5 GDP/GTP exchange factor|Rab5ef|Rabaptin 5 associated exchange factor for Rab5|Rabaptin-5-associated exchange factor for Rab5|Rabex 5|Rabex-5|RABGEF 1|Rabgef1|RABX 5|RABX5|RABX5_HUMAN|RAP1|Rin 2|Rin2
- RAB guanine nucleotide exchange factor (GEF) 1|Rab5 GDP/GTP exchange factor|Rab5ef|Rabaptin 5 associated exchange factor for Rab5|Rabaptin-5-associated exchange factor for Rab5|Rabex 5|Rabex-5|RABGEF 1|Rabgef1|RABX 5|RABX5|RABX5_HUMAN|RAP1|Rin 2|Rin2
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9UJ41
- Q9UJ41
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice