Anti-TULP3 Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: TULP3
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77811-729
- 77803-729
- Antigen Symbol:
- TULP3
- TULP3
- Antigen Name:
- Tubby-related protein 3 protein (1-260AA)
- Tubby-related protein 3 protein (1-260AA)
- Antigen Synonyms:
- MGC45295|Tubby like protein 3|Tubby related protein 3|Tubby-like protein 3|Tubby-related protein 3|TUBL 3|TUBL3|TULP 3|TULP3|TULP3_HUMAN
- MGC45295|Tubby like protein 3|Tubby related protein 3|Tubby-like protein 3|Tubby-related protein 3|TUBL 3|TUBL3|TULP 3|TULP3|TULP3_HUMAN
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- O75386
- O75386
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on Blue ice
- Shipped on Blue ice
Specifications
Frequently Bought Together