Anti-USP26 Rabbit Recombinant Polyclonal Antibody
Catalog # 77839-206
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Name:Ubiquitin carboxyl-terminal hydrolase 26 protein (664-913AA)
- Antigen Symbol:USP26
- Clonality:Recombinant Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Mouse
- Western Blot:Yes
- Size:50 µl
- Form:Liquid
- Gene ID:Q9BXU7
- Antigen Synonyms:Deubiquitinating enzyme 26| MGC120066| MGC120067| MGC120068| Ubiquitin carboxyl terminal hydrolase 26| Ubiquitin carboxyl-terminal hydrolase 26| Ubiquitin specific peptidase 26| Ubiquitin specific processing protease 26| Ubiquitin specific protease 26| Ubiquitin thioesterase 26| Ubiquitin thiolesterase 26| Ubiquitin-specific-processing protease 26| UBP26_HUMAN| USP 26| USP26
- Storage Buffer:PBS, 0.02% sodium azide, 50% glycerol, pH 7.3
- Storage Temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on Blue ice
- Purification:Antigen affinity purified
- Packaging:Vial
Specifications
About this item
Recommended dilution: WB: 1:1000 to 1:5000, IHC: 1:20 to 1:200
Type:
Antigen: USP26
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse