Anti-RHOQ Rabbit Polyclonal Antibody
Catalog # 77797-323
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Name:TC10A
- Antigen Symbol:RHOQ
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Mouse,Rat
- Western Blot:Yes
- Size:100 µl
- Form:Liquid
- Gene ID:P17081
- Antigen Synonyms:ARHQ|Ras homolog gene family member Q|RAS like family 7 member A|Ras like protein family member 7A|Ras like protein TC10|Ras related GTP binding protein TC10|Ras-like protein family member 7A|Ras-like protein TC10|RASL7A|Rho related GTP binding protein RhoQ|Rho-related GTP-binding protein RhoQ|Rhoq|RHOQ_HUMAN|TC10A
- Storage Buffer:PBS, 0.1% sodium azide, 50% glycerol, pH 7.3
- Storage Temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Purification:Antigen affinity purified
- Packaging:Vial
Specifications
About this item
Type:
Antigen: RHOQ
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat