Anti-CCDC32 Rabbit Recombinant Polyclonal Antibody (FITC)
New Product
Type:
Antigen: CCDC32
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77841-704
- 77841-705
- Antigen Symbol:
- CCDC32
- CCDC32
- Antigen Name:
- Coiled-coil domain-containing protein 32 protein (1-185AA)
- Coiled-coil domain-containing protein 32 protein (1-185AA)
- Antigen Synonyms:
- CCDC32| C15orf57Coiled-coil domain-containing protein 32
- CCDC32| C15orf57Coiled-coil domain-containing protein 32
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9BV29
- Q9BV29
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on Blue ice
- Shipped on Blue ice