Anti-MPZL3 Rabbit Recombinant Polyclonal Antibody
New Product
Recommended dilution: IF: 1:50 to 1:200
Type:
Antigen: MPZL3
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77829-444
- 77829-443
- Antigen Symbol:
- MPZL3
- MPZL3
- Antigen Name:
- Myelin protein zero-like protein 3 protein (32-158AA)
- Myelin protein zero-like protein 3 protein (32-158AA)
- Antigen Synonyms:
- MPZL3_HUMAN|Myelin protein zero like 3|Myelin protein zero-like protein 3|PRO7425|UNQ2966
- MPZL3_HUMAN|Myelin protein zero like 3|Myelin protein zero-like protein 3|PRO7425|UNQ2966
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q6UWV2
- Q6UWV2
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice