Anti-GNPNAT1 Rabbit Recombinant Polyclonal Antibody (FITC)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: GNPNAT1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77834-056
- 77834-055
- Antigen Symbol:
- GNPNAT1
- GNPNAT1
- Antigen Name:
- Glucosamine 6-phosphate N-acetyltransferase protein (1-184AA)
- Glucosamine 6-phosphate N-acetyltransferase protein (1-184AA)
- Antigen Synonyms:
- AU017428|AU040593|EMeg32|FLJ10607|Glucosamine 6 phosphate acetyltransferase|Glucosamine 6 phosphate N acetyltransferase|Glucosamine 6-phosphate N-acetyltransferase|Glucosamine phosphate N acetyltransferase 1|GNA1|GNA1_HUMAN|GNPNAT|GNPNAT1|Gpnat1|Gsnpat|Phosphoglucosamine acetylase|Phosphoglucosamine transacetylase|RGD1563144
- AU017428|AU040593|EMeg32|FLJ10607|Glucosamine 6 phosphate acetyltransferase|Glucosamine 6 phosphate N acetyltransferase|Glucosamine 6-phosphate N-acetyltransferase|Glucosamine phosphate N acetyltransferase 1|GNA1|GNA1_HUMAN|GNPNAT|GNPNAT1|Gpnat1|Gsnpat|Phosphoglucosamine acetylase|Phosphoglucosamine transacetylase|RGD1563144
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q96EK6
- Q96EK6
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together