Anti-MOV10L1 Rabbit Recombinant Antibody (Biotinylated)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: MOV10L1
Clonality: Recombinant
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77838-101
- 77838-102
- Antigen Symbol:
- MOV10L1
- MOV10L1
- Antigen Name:
- RNA helicase Mov10l1 protein (336-425AA)
- RNA helicase Mov10l1 protein (336-425AA)
- Antigen Synonyms:
- Cardiac helicase activated by MEF2 protein|Cardiac specific RNA helicase|CHAMP|Csm|M10L1_HUMAN|Moloney leukemia virus 10 like protein 1|Moloney leukemia virus 10-like protein 1|MOV10 like 1|MOV10 like protein 1|MOV10-like protein 1|MOV10L1|Moloney leukemia virus 10 like 1|homolog (mouse)|Putative helicase Mov10l1
- Cardiac helicase activated by MEF2 protein|Cardiac specific RNA helicase|CHAMP|Csm|M10L1_HUMAN|Moloney leukemia virus 10 like protein 1|Moloney leukemia virus 10-like protein 1|MOV10 like 1|MOV10 like protein 1|MOV10-like protein 1|MOV10L1|Moloney leukemia virus 10 like 1|homolog (mouse)|Putative helicase Mov10l1
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9BXT6
- Q9BXT6
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice