Anti-LAPTM5 Rabbit Recombinant Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: IHC:1:20 to 1:200
Type:
Antigen: LAPTM5
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77824-773
- 77824-772
- Antigen Symbol:
- LAPTM5;KIAA0085;Lysosomal-associated transmembrane protein 5;Lysosomal-associated multitransmembrane protein 5;Retinoic acid-inducible E3 protein
- LAPTM5;KIAA0085;Lysosomal-associated transmembrane protein 5;Lysosomal-associated multitransmembrane protein 5;Retinoic acid-inducible E3 protein
- Antigen Name:
- Lysosomal-associated transmembrane protein 5 protein (205-262AA)
- Lysosomal-associated transmembrane protein 5 protein (205-262AA)
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q13571
- Q13571
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice