Anti-PAFAH1B3 Rabbit Recombinant Polyclonal Antibody (FITC)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: PAFAH1B3
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77824-031
- 77824-030
- Antigen Symbol:
- PAFAH1B3
- PAFAH1B3
- Antigen Name:
- Platelet-activating factor acetylhydrolase IB subunit gamma protein (103-187AA)
- Platelet-activating factor acetylhydrolase IB subunit gamma protein (103-187AA)
- Antigen Synonyms:
- PA1B3_HUMAN| PAF acetylhydrolase 29 kDa subunit| PAF AH 29 kDa subunit| PAF AH subunit gamma| PAF-AH 29 kDa subunit| PAF-AH subunit gamma| PAFAH subunit gamma| Pafah1b3| PAFAHG| Platelet activating factor acetylhydrolase IB subunit gamma| Platelet activating factor acetylhydrolase isoform Ib gamma subunit (29kD)| Platelet activating factor acetylhydrolase isoform Ib gamma subunit 29kDa| Platelet activating factor acetylhydrolase isoform Ib gamma subunit| Platelet-activating factor acetylhydrolase IB subunit gamma
- PA1B3_HUMAN| PAF acetylhydrolase 29 kDa subunit| PAF AH 29 kDa subunit| PAF AH subunit gamma| PAF-AH 29 kDa subunit| PAF-AH subunit gamma| PAFAH subunit gamma| Pafah1b3| PAFAHG| Platelet activating factor acetylhydrolase IB subunit gamma| Platelet activating factor acetylhydrolase isoform Ib gamma subunit (29kD)| Platelet activating factor acetylhydrolase isoform Ib gamma subunit 29kDa| Platelet activating factor acetylhydrolase isoform Ib gamma subunit| Platelet-activating factor acetylhydrolase IB subunit gamma
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q15102
- Q15102
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on Blue ice
- Shipped on Blue ice