Anti-USP5 Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: USP5
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77804-163
- 77804-164
- Antigen Symbol:
- USP5
- USP5
- Antigen Name:
- Ubiquitin carboxyl-terminal hydrolase 5 protein (201-500AA)
- Ubiquitin carboxyl-terminal hydrolase 5 protein (201-500AA)
- Antigen Synonyms:
- Deubiquitinating enzyme 5|Isopeptidase T|ISOT 1|ISOT|Ubiquitin carboxyl-terminal hydrolase 5|Ubiquitin isopeptidase T|Ubiquitin specific peptidase 5 (isopeptidase T)|Ubiquitin specific peptidase 5|Ubiquitin specific processing protease 5|Ubiquitin specific protease 5 (isopeptidase T)|Ubiquitin specific protease 5 (ubiquitin isopeptidase T)|Ubiquitin thioesterase 5|Ubiquitin thiolesterase 5|Ubiquitin-specific-processing protease 5|UBP5_HUMAN|Usp5
- Deubiquitinating enzyme 5|Isopeptidase T|ISOT 1|ISOT|Ubiquitin carboxyl-terminal hydrolase 5|Ubiquitin isopeptidase T|Ubiquitin specific peptidase 5 (isopeptidase T)|Ubiquitin specific peptidase 5|Ubiquitin specific processing protease 5|Ubiquitin specific protease 5 (isopeptidase T)|Ubiquitin specific protease 5 (ubiquitin isopeptidase T)|Ubiquitin thioesterase 5|Ubiquitin thiolesterase 5|Ubiquitin-specific-processing protease 5|UBP5_HUMAN|Usp5
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P45974
- P45974
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together