Anti-PIP Rabbit Recombinant Polyclonal Antibody
New Product
Recommended dilution: IHC: 1:20 to 1:200
Type:
Antigen: PIP
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77794-182
- 77794-183
- Antigen Symbol:
- PIP
- PIP
- Antigen Name:
- Prolactin-inducible protein (29-146AA)
- Prolactin-inducible protein (29-146AA)
- Antigen Synonyms:
- BRST-2|BRST2|GCDFP-15|GCDGP15|gp17|GPIP4|Gross cystic disease fluid protein 15|PIP|PIP_HUMAN|Prolactin induced protein|Prolactin inducible protein precursor|Prolactin-induced protein|Prolactin-inducible protein|SABP|Secretory actin binding protein|Secretory actin-binding protein
- BRST-2|BRST2|GCDFP-15|GCDGP15|gp17|GPIP4|Gross cystic disease fluid protein 15|PIP|PIP_HUMAN|Prolactin induced protein|Prolactin inducible protein precursor|Prolactin-induced protein|Prolactin-inducible protein|SABP|Secretory actin binding protein|Secretory actin-binding protein
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P12273
- P12273
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice