Anti-MSTO1 Rabbit Recombinant Polyclonal Antibody (HRP)
New Product
Type:
Antigen: MSTO1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77840-285
- 77840-284
- Antigen Symbol:
- MSTO1
- MSTO1
- Antigen Name:
- Protein misato homolog 1 protein (1-280AA)
- Protein misato homolog 1 protein (1-280AA)
- Antigen Synonyms:
- MSTO1|LST005|SLTP005|Protein misato homolog 1
- MSTO1|LST005|SLTP005|Protein misato homolog 1
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- HRP (Horseradish Peroxidase)
- HRP (Horseradish Peroxidase)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9BUK6
- Q9BUK6
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Frequently Bought Together