Anti-ROMO1 Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: ROMO1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77797-700
- 77797-699
- Antigen Symbol:
- ROMO1
- ROMO1
- Antigen Name:
- Reactive oxygen species modulator 1 protein (45-79AA)
- Reactive oxygen species modulator 1 protein (45-79AA)
- Antigen Synonyms:
- bA353C18.2|C20orf52|Mitochondrial targeting GXXXG protein|MTGMP|Protein MGR2 homolog|Reactive oxygen species modulator 1|ROMO1|ROMO1_HUMAN|ROS modulator 1
- bA353C18.2|C20orf52|Mitochondrial targeting GXXXG protein|MTGMP|Protein MGR2 homolog|Reactive oxygen species modulator 1|ROMO1|ROMO1_HUMAN|ROS modulator 1
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P60602
- P60602
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice