Anti-PRPF8 Rabbit Recombinant Antibody (HRP (Horseradish Peroxidase))
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: PRPF8
Clonality: Recombinant
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77829-693
- 77829-692
- Antigen Symbol:
- PRPF8
- PRPF8
- Antigen Name:
- Pre-mRNA-processing-splicing factor 8 protein (2037-2170AA)
- Pre-mRNA-processing-splicing factor 8 protein (2037-2170AA)
- Antigen Synonyms:
- 220 kDa U5 snRNP specific protein|220 kDa U5 snRNP-specific protein|Apoptosis regulated protein 1|Apoptosis regulated protein 2|HPRP8|p220|Pre mRNA processing factor 8|Pre mRNA-processing factor 8|S. cerevisiae|homolog of|Pre-mRNA-processing-splicing factor 8|Precursor mRNA processing protein|PRP8|PRP8 homolog|PRP8 pre mRNA processing factor 8 homolog|PRP8_HUMAN|PRPC8|Prpf8|Retinitis pigmentosa 13 (autosomal dominant)|RP13|SNRNP220|Splicing factor Prp8|U5 snRNP specific protein|U5 snRNP specific protein (220 kD)|ortholog of S. cerevisiae Prp8p
- 220 kDa U5 snRNP specific protein|220 kDa U5 snRNP-specific protein|Apoptosis regulated protein 1|Apoptosis regulated protein 2|HPRP8|p220|Pre mRNA processing factor 8|Pre mRNA-processing factor 8|S. cerevisiae|homolog of|Pre-mRNA-processing-splicing factor 8|Precursor mRNA processing protein|PRP8|PRP8 homolog|PRP8 pre mRNA processing factor 8 homolog|PRP8_HUMAN|PRPC8|Prpf8|Retinitis pigmentosa 13 (autosomal dominant)|RP13|SNRNP220|Splicing factor Prp8|U5 snRNP specific protein|U5 snRNP specific protein (220 kD)|ortholog of S. cerevisiae Prp8p
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- HRP (Horseradish Peroxidase)
- HRP (Horseradish Peroxidase)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q6P2Q9
- Q6P2Q9
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together